Learn More
Sulfuric Acid, 10% v/v with 1% w/v Manganese Sulfate, Reagents
Specifications
Specifications
| pH | <1 |
| Color | Light Pink |
| Boiling Point | 100°C |
| Packaging | Poly Bottle |
| Specific Gravity | 1.11 |
| Quantity | 1 L |
| Physical Form | Liquid |
| Percent Purity | 10% |
| Grade | Reagent |
| Chemical Name or Material | Sulfuric Acid, 10% v/v with 1% w/v Manganese Sulfate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.