Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid (ACS Grade), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
2.5 L
6.5 gal.
6 x 2.5 L
225 lb.
700 lb.
Packaging:
Glass Bottle
PPQ Resin Bottle
Poly Bottle
Poly Drum
Safe-Cote™ Glass Bottle
Specifications
Specifications
| pH | 0.3 |
| Color | Colorless to Brown |
| Melting Point | 10°C |
| Boiling Point | 290°C |
| Density | 1.84g/cm³ |
| Heavy Metals (as Pb) | 0.8ppm max. |
| Identification | Pass Test |
| Packaging | Poly Bottle |
| Residue after Ignition | 5 ppm max. |
| Vapor Pressure | <0.001 mmHg at 20°C |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Causes severe burns by all exposure routes. Reacts violently with water. Contact with combustible material may cause fire. Exposure to strong inorganic mists containing sulfuric acid may cause cancer by inhalation. Hygroscopic. Use personal protective equipment. Do not allow contact with water. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. NFPA Health:3 Flammability:0 Instability:2 |
| DOTInformation | DOT Class 8, : Corrosive |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction