Learn More
Sulfuric Acid, FCC, 95-98%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Sulfuric Acid, FCC is considered a general purpose food additive and is used to directly control pH during the processing of food by discouraging the growth of bacteria and spoilage microbes. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Percent Purity | 95 to 98% |
| Grade | FCC |
| Chemical Name or Material | Sulfuric Acid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.