Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sulfuric Acid, Concentrated, Reagent Grade
SCI_ED

Catalog No. s25894 Shop All Fisherbrand Products
Click to view available options
Quantity:
2.5 L
Packaging:
Reagent Grade

Catalog No. S25894

Only null left
Add to Cart
Edge
Add to Cart

Ideal for educational use in classrooms, labs, and training environments.

  • Product is for educational use
  • Available in a 2.5 L size

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Density 1.840
Packaging Reagent Grade
Quantity 2.5 L
Physical Form Liquid
Linear Formula H2SO4
Formula Weight 98.07
Percent Purity 95 to 98%
Grade ACS
Chemical Name or Material Sulfuric Acid, Concentrated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.