Learn More
Sulfuric Acid-Silver Sulfate Reagent, for Chemical Oxygen Demand (COD), Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Packaging | Safety-coated Glass Bottle |
| CAS Min % | 97.0, 0.53 |
| CAS Max % | 100.0, 0.55 |
| Physical Form | Liquid |
| Color | Colorless |
| Quantity | 16 oz. |
| Chemical Name or Material | Sulfuric Acid-Silver Sulfate Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.