Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid-Silver Sulfate Reagent, for Chemical Oxygen Demand (COD), Ricca Chemical
SDP

Catalog No. 835032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
16 oz.
1 L
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8350-32 1 L Safety-coated Glass Bottle
83504 120 mL Amber Glass Bottle
8350-16 16 oz. Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 8350-32 Supplier Ricca Chemical Company Supplier No. 835032
Add to Cart
Add to Cart

For Analysis of Chemical Oxygen Demand (COD)

Chemical Identifiers

CAS 7664-93-9 , 10294-26-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Packaging Safety-coated Glass Bottle
CAS Min % 97.0, 0.53
CAS Max % 100.0, 0.55
Physical Form Liquid
Color Colorless
Quantity 1 L
Chemical Name or Material Sulfuric Acid-Silver Sulfate Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.