Sulphur Precipitated, 0.995, Alfa Aesar™

Manufacturer: Alfa Aesar 10785A3

Quantity 2kg

 View more versions of this product

Catalog No. AA10785A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7704-34-9
    Physical Form Precipitated
    Assay Percent Range 99.50%
    MDL Number MFCD00085316
    Chemical Name or Material Sulphur

    Specifications

    Quantity 2kg
    Color Pale yellow
    DOT Information DOT Class: 4.1, Packaging Group: III
    EINECS Number 231-722-6
    Physical Form Precipitated
    Ignition Point 248°C (478°F)
    Odor Weak
    Assay Percent Range 99.50%
    MDL Number MFCD00085316
    CAS 7704-34-9
    Health Hazard 1 H228-H315; Flammable solid. Causes skin irritation.
    Chemical Name or Material Sulphur

    H228-H315; Flammable solid. Causes skin irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit