Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Sumanirole maleate
GREENER_CHOICE

Catalog No. p-4784619
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective D2 agonist

High affinity D2 receptor agonist (EC50 values are between 17 and 75 nM in cell-based assays). Originally reported to exhibit >200-fold selectivity for the D2 receptor versus other dopamine receptor subtypes (Ki values are 9.0, 1940, >2190 and >7140 for D2, D3, D4 and D1 receptors respectively). More recently reported to be around 32-fold selective for D2 over D3 receptors (Ki values are 17.1 and 546 nM respectively). Exhibits antiParkinsonian activity.

Chemical Identifiers

CAS 179386-44-8
Molecular Formula C15H17N3O5

Specifications

Quantity 10 mg
Formula Weight 319.31
Purity >98%
Molecular Weight (g/mol) 319.317
Chemical Name or Material Sumanirole maleate
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.