Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO Activating Enzyme E1 (SAE1) Antibody (9D8), Novus Biologicals™
SDP

Catalog No. NB298304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298304 50 μL
NB298305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298304 Supplier Novus Biologicals Supplier No. NBP32633650UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SUMO Activating Enzyme E1 (SAE1) Monoclonal antibody specifically detects SUMO Activating Enzyme E1 (SAE1) in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry

Specifications

Antigen SUMO Activating Enzyme E1 (SAE1)
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 9D8
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A
Host Species Rabbit
Immunogen A synthesized peptide derived from Human SUMO Activating Enzyme E1 (SAE1) [UniProt Q9UBE0]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10055
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.