Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO Activating Enzyme E1 (SAE1) Antibody (OTI6B11), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP274410A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274410A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274410A32 Supplier Novus Biologicals Supplier No. NBP274410AF532

Mouse Monoclonal Antibody

SUMO Activating Enzyme E1 (SAE1) Monoclonal specifically detects SUMO Activating Enzyme E1 (SAE1) in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SUMO Activating Enzyme E1 (SAE1)
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6B11
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A
Gene Symbols SAE1
Host Species Mouse
Immunogen Full length recombinant protein of human SUMO Activating Enzyme E1 (SAE1) (NP_005491) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10055
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.