Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO Activating Enzyme E1 (SAE1) Antibody (OTI6B11), Novus Biologicals™
SDP

Catalog No. NBP245683 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP245683 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP245683 Supplier Novus Biologicals Supplier No. NBP245683

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunohistochemistry

SUMO Activating Enzyme E1 (SAE1) Monoclonal specifically detects SUMO Activating Enzyme E1 (SAE1) in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SUMO Activating Enzyme E1 (SAE1)
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6B11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:150
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A
Gene Symbols SAE1
Host Species Mouse
Immunogen Full length human recombinant protein of human SAE1 (NP_005491) produced in HEK293T cell.
Molecular Weight of Antigen 38.3 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10055
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.