Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUMO-interacting Motif (SIM) Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23070025UL
Description
SUMO-interacting Motif (SIM) Polyclonal specifically detects SUMO-interacting Motif (SIM) in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SUMO-interacting Motif (SIM) | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
Q8NDZ2 | |
SIMC1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MKIQQLHPANAKTVEWDWKLLTYVMEEEGQTLPGRVLFLRYVVQTLEDDFQQTLRRQRQHLQQSIANMVLSCDKQPHNVRDVIKWLVKA | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Chromosome 5 Open Reading Frame 25, Oocyte Maturation Associated 1, OOMA1, Platform Element For Inhibition Of Autolytic Degradation, PLEIAD, SIMC1, SUMO-Interacting Motif-Containing Protein 1, SUMO-Interacting Motifs Containing 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
375484 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction