Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUMO-interacting Motif (SIM) Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SUMO-interacting Motif (SIM) |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SUMO-interacting Motif (SIM) Polyclonal specifically detects SUMO-interacting Motif (SIM) in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SUMO-interacting Motif (SIM) | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Chromosome 5 Open Reading Frame 25, Oocyte Maturation Associated 1, OOMA1, Platform Element For Inhibition Of Autolytic Degradation, PLEIAD, SIMC1, SUMO-Interacting Motif-Containing Protein 1, SUMO-Interacting Motifs Containing 1 | |
SIMC1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
Polyclonal | |
Rabbit | |
Human | |
Q8NDZ2 | |
375484 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MKIQQLHPANAKTVEWDWKLLTYVMEEEGQTLPGRVLFLRYVVQTLEDDFQQTLRRQRQHLQQSIANMVLSCDKQPHNVRDVIKWLVKA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title