Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO2/3 Antibody (SPM572) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB397215 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397215 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397215 Supplier Novus Biologicals Supplier No. NBP2344310.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SUMO2/3 Monoclonal antibody specifically detects SUMO2/3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Proximity Ligation Assay, CyTOF

Specifications

Antigen SUMO2/3
Applications Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Proximity Ligation Assay, CyTOF
Classification Monoclonal
Clone SPM572
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5 to 1 μg/mL, Immunohistochemistry-Paraffin 0.5 to 1 μg/mL, Proximity Ligation Assay, CyTOF-ready
Formulation 10 mM PBS
Gene Alias HSMT3, small ubiquitin-like modifier 2, SMT3A, SMT3B, SMT3H2, SUMO2, SUMO3
Host Species Mouse
Immunogen Recombinant human SUMO2/3 protein
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 6613
Target Species Human
Content And Storage Store at -20°C to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.