Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SUN 11602
Click to view available options
Quantity:
10 mg
50 mg
Description
SUN 11602 is a basic fibroblast growth factor (bFGF) mimetic. Mimics the neuroprotective effects of bFGF; prevents glutamate-induced neuronal death. Increases levels of CALB1 gene expression in cerebrocortical neurons. Triggers phosphorylation of FGFR, ERK1 and ERK2.
Specifications
Specifications
Purity | >98% |
Content And Storage | -20°C |
Quantity | 10 mg |
Product Type | SUN 11602 |
Molecular Weight (g/mol) | 451.61 |
Inhibitors | SUN 11602 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction