Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SUN-B 8155
GREENER_CHOICE

Catalog No. 255010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-501-0 10 mg
25-505-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-501-0 Supplier Tocris Bioscience™ Supplier No. 2550/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-peptide calcitonin (CT) receptor agonist

Non-peptide calcitonin (CT) receptor agonist that fully stimulates cAMP production in CHO/hCTR cells (EC50 = 21 μM). Causes a significant reduction in serum calcium concentrations in vivo following i.p. administration.

Chemical Identifiers

CAS 345893-91-6
Molecular Formula C14H15N3O3

Specifications

Quantity 10 mg
Formula Weight 273.29
Purity >98%
Molecular Weight (g/mol) 273.292
Chemical Name or Material SUN-B 8155
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.