Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SUN-B 8155
GREENER_CHOICE

Catalog No. p-4784621
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-peptide calcitonin (CT) receptor agonist

Non-peptide calcitonin (CT) receptor agonist that fully stimulates cAMP production in CHO/hCTR cells (EC50 = 21 μM). Causes a significant reduction in serum calcium concentrations in vivo following i.p. administration.

Chemical Identifiers

CAS 345893-91-6
Molecular Formula C14H15N3O3

Specifications

Quantity 10 mg
Formula Weight 273.29
Purity >98%
Molecular Weight (g/mol) 273.292
Chemical Name or Material SUN-B 8155
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.