Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUN1 Antibody (12.10F), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP259943R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-59943R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-59943R Supplier Novus Biologicals Supplier No. NBP259943R

Mouse Monoclonal Antibody

SUN1 Monoclonal specifically detects SUN1 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Knockdown Validated.

Specifications

Antigen SUN1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Monoclonal
Clone 12.10F
Conjugate DyLight 550
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias KIAA0810UNC84AFLJ12407, MGC176649, Protein unc-84 homolog A, Sad1 and UNC84 domain containing 1, Sad1 unc-84 domain protein 1, Sad1/unc-84 protein-like 1, SUN domain-containing protein 1, unc-84 homolog A, unc-84 homolog A (C. elegans)
Gene Symbols SUN1
Host Species Mouse
Immunogen GST fused to residues encoded by exons 6, 8, 10
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline modENCODE Antibodies
Primary or Secondary Primary
Gene ID (Entrez) 23353
Test Specificity Specificity of mouse, rat SUN1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.