Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUN1 Antibody (HL1946) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294916 Supplier Novus Biologicals Supplier No. NBP325701
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SUN1 Monoclonal antibody specifically detects SUN1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SUN1
Applications Immunocytochemistry
Classification Monoclonal
Clone HL1946
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias KIAA0810UNC84AFLJ12407, MGC176649, Protein unc-84 homolog A, Sad1 and UNC84 domain containing 1, Sad1 unc-84 domain protein 1, Sad1/unc-84 protein-like 1, SUN domain-containing protein 1, unc-84 homolog A, unc-84 homolog A (C. elegans)
Host Species Rabbit
Immunogen Recombinant fragment of human SUN1.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline modENCODE Antibodies
Primary or Secondary Primary
Gene ID (Entrez) 23353
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.