Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SUN2 |
---|---|
Concentration | 0.43 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5699
![]() ![]() |
Proteintech Group Inc
275561AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
UNC84B may form a physical interaction between the nuclear envelope and the centrosome.Specifications
SUN2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8BJS4, Q9UH99 | |
223697, 25777 | |
SUN2 Fusion Protein Ag16077 | |
Primary | |
-20°C | |
SUN2 |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FRIGG, KIAA0668, Protein unc 84 homolog B, Rab5 interacting protein, Rab5IP, Sad1/unc 84 protein like 2, SUN2, unc 84 homolog B (C. elegans), UNC84B | |
Sun2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title