Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUOX Antibody (OTI5A2), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP274411C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274411C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274411C Supplier Novus Biologicals Supplier No. NBP274411C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SUOX Monoclonal specifically detects SUOX in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SUOX
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5A2
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 1.8.3.1, sulfite oxidase, sulfite oxidase, mitochondrial
Gene Symbols SUOX
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 320-545 of human SUOX (NP_000447) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6821.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.