Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supel™ Swift HLB SPE Tubes, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Model:
200 mg (bed)
30 mg (bed)
60 mg (bed)
Description
- This phase is ideal for extracting a broad range of compounds from aqueous samples, including food and environmental samples, as well as biological samples (eg. serum and plasma).
- Because of the hydrophilic and lipophilic balanced (HLB) property, the polymer material can selectively retain a wide range of compounds with different polarities and LogP values.
- Traditional HLB SPE uses a five-step process for sample preparation that normally includes the conditioning of sorbents with an organic solvent followed by an equilibration step to return sorbents to sample loading conditions. The product design allows for direct sample loading onto the sorbent bed, thereby eliminating conditioning of the cartridge. This product feature can both save time and simplify sample preparation, which may also serve to reduce sample processing errors.
Specifications
Specifications
| Type | 80 to 210 Å Pore Size |
| Includes | 40-75 μm (>90%) particle size |
| Model | 60 mg (bed) |
| Volume (Metric) | 3 mL |
| Description | Detection MS friendly |
Supel is a trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction