Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ SUPELCOGEL™ Pb, 6% Crosslinked HPLC Column, 9 μm Particle Size

Catalog No. 111003025 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
10 cm
30 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-100-3025 30 cm
11-100-3107 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-3025 Supplier MilliporeSigma™ Supelco™ Supplier No. 59343
Add to Cart
Add to Cart

SUPELCOGEL™ Pb, 6% crosslinked HPLC column may be used for the separation and determination of d-mannose in human serum samples using liquid chromatography coupled with tandem mass spectrometry.

SUPELCOGEL™ Pb, 6% Crosslinked HPLC Column has been used for the separation of:. D-mannose in human serum samples during its determination using liquid chromatography-tandem mass spectrometry (LC-MS/MS) equipped with electrospray ionization (ESI) source and selected reaction monitoring (SRM) detection. Free sugars in switchgrass extractives during their quantification using high performance liquid chromatography equipped with refractive index (HPLC-RI) detector.. Total hemicellulose and cellulose sugars in sugarcane straw hydrolyzate during their determination using HPLC equipped with infra-red (IR) detector.
TRUSTED_SUSTAINABILITY

Specifications

Crosslinking 0.06
Diameter (Metric) 7.8 mm
Length (Metric) 30 cm
Particle Size 9 μm
Stationary Phase Sulfonated Polystyrene
Product Line SUPELCOGEL
Matrix Sulfonated Polystyrene/Divinylbenzene Phase/Spherical Particle Platform
Phase Ion Exclusion
Technique Crosslinked HPLC

SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.