Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Super AquaBlue ELISA Substrate
Click to view available options
Quantity:
100 mL
500 mL
Description
- Highly sensitive and extremely stable (18 months at room temperature or 24 months at 4°C).
- Engineered for convenience as a single component (no additional H2O2 or other components are necessary)
- Suitable for endpoint reactions, but is also ideal for kinetic assays; stopping the reaction with acid stop solution does not change the color or absorption properties of the blue-green end product
Specifications
Specifications
| Product Type | ELISA Substrate |
| Quantity | 100 mL |
| Detection Method | Colorimetric |
| Substrate Properties | Chemical Substrate |
| Substrate Type | HRP (Horseradish Peroxidase) |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction