Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ SuperBlock™ Blocking Buffer, blotting
Click to view available options
Buffer:
PBS
TBS
Description
Thermo Scientific SuperBlock Blocking Buffer, blotting is an optimized PBS solution containing a protein that provides incredibly fast and effective blocking for western blot analysis. Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.
Features of SuperBlock Blocking Buffer, blotting include:
• Fast—block membranes in 5 to 10 minutes
• Flexible—guaranteed to be biotin-free for use with streptavidin system
• Low background—the non-serum protein solution yields a high signal-to-noise ratio
• Stable—store buffer at 4°C for one year; store blocked plates dry for up to 12 months
Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.
• Fast—block membranes in 5 to 10 minutes
• Flexible—guaranteed to be biotin-free for use with streptavidin system
• Low background—the non-serum protein solution yields a high signal-to-noise ratio
• Stable—store buffer at 4°C for one year; store blocked plates dry for up to 12 months
Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.

Specifications
Specifications
Description | SuperBlock (TBS) Blocking Buffer, blotting |
Product Type | Blocking Buffer |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | Western Blot |
Product Line | SuperBlock™ |
Concentration | 1X |
Buffer | TBS |
Blocking Agent | Single purified protein |
Content And Storage | Upon receipt store at 4°C. Product is shipped at ambient temperature. |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction