Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ IV Template Switching RT Master Mix

Catalog No. p-200009447
Encompass_Preferred
Click to view available options
No. of Reactions:
48 Reactions
5 x 96 Reactions
96 Reactions

Invitrogen SuperScript IV Template-Switching RT Master Mix is designed for high template switching efficiency in cDNA synthesis reactions.

Invitrogen SuperScript IV Template-Switching RT Master Mix is designed for high template switching efficiency in cDNA synthesis reactions. It contains SuperScript IV Reverse Transcriptase, RNase Inhibitor, and all other reverse transcription reaction components in a convenient premixed format. Only RNA template, RT primer, and template switching oligo (TSO) of choice need to be added.

Features of SuperScript IV Template-Switching RT Master Mix include:
• Formulated for high quality full-length cDNA synthesis
• Includes proprietary enhancer to maximize template switching efficiency
• Exceptional sensitivity for easy detection of low abundance targets (as low as single cell or 2 pg total RNA)
• Cell lysis buffer included for direct cDNA synthesis without RNA purification

Procedure overview
To maximize template switching efficiency, SuperScript IV Template-Switching RT Master Mix contains proprietary template switching enhancer and leverages the terminal deoxynucleotidyl transferase (TdT) activity of SuperScript IV Reverse Transcriptase. When the reverse transcriptase reaches the 5’ end of the RNA template, TdT activity adds 1–3 extra nucleotides to the cDNA end, enabling binding of the template switching oligo and incorporation of known sequence at the 3’ end of the resulting cDNA. This cDNA can be used for amplification by PCR, gene expression analysis by real-time PCR, or as a template for 5’RACE, RNA-seq, or second strand cDNA synthesis.This master mix is compatible with a wide input range of total RNA (from 2 pg to 4 µg) and is supplied with lysis buffer to enable direct cDNA synthesis from mammalian cells (1–1,000 cells per sample).

Related products
SuperScript IV Template-Switching RT Master Mix is compatible with various RT primers and self-designed template switching oligos. We offer Template Switching Oligo (Cat. No. A65427) and Capturing Oligo (dT) Primer (A65429), both containing the same adaptor sequence complementary to Preamplification Primer (A65428). Alternatively, a full set of reagents for template switching cDNA synthesis and preamplification is available in the SuperScript IV Single Cell/Low-Input cDNA PreAmp Kit (11752048).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 4X
Content And Storage • 4X SuperScript IV cDNA Synthesis Master Mix (240 μL)
• 10X Lysis Buffer (48 μL)
• Nuclease-free water (1.25 mL)

Store at -15°C to -25°C.

Format Master Mix
GC-Rich PCR Performance High
Reaction Speed 30 min.
Technique Template Switching Reverse Transcription
Quantity 48 reactions
Final Product Type First-Strand cDNA
No. of Reactions 48 Reactions
Reagent Type Reverse Transcription
Starting Material Cells or RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.