Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript IV UniPrime One-Step RT-PCR System

Catalog No. p-7229280
Encompass_Preferred
Click to view available options
Description:
Colored
Dye-free
No. of Reactions:
100 x 50 μL Reactions
25 x 50 μL Reactions
500 x 50 μL Reactions
This item is not returnable. View return policy

Catalog No. 12-596-025

Supplier: Invitrogen™ 12596025

Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Invitrogen SuperScript IV UniPrime One-Step RT-PCR System is a convenient one-step formulation for performing fast cDNA synthesis and PCR amplification in a single tube.

Invitrogen SuperScript IV UniPrime One-Step RT-PCR System is a convenient one-step formulation for performing fast cDNA synthesis and PCR amplification in a single tube. It combines the high-processivity SuperScript IV Reverse Transcriptase (RT) with a novel UniPrime RT-PCR Master Mix to provide easier use and unmatched performance when compared to other one-step RT-PCR kits.

The Invitrogen SuperScript IV UniPrime One-Step RT-PCR System offers a convenient one-step process  for performing fast cDNA synthesis and PCR amplification in a single tube. It combines high-processivity SuperScript IV Reverse Transcriptase (RT) with a novel UniPrime RT-PCR master mix for easier use and unmatched  performance compared to other one-step RT-PCR kits.

Highlights
Simplified workflow—universal primer annealing at 60°C decreases optimization time
Easy pipetting—colored buffers for visual tracking of reaction setup and direct sample loading on gels
Suitable for automation—two-phase hot-start activation mechanism for room temperature setup and benchtop stability for up to 24 hours
Superior performance—high yields and reliable target detection even with challenging RNA samples

Combination of superior enzymes
The SuperScript IV UniPrime One-Step RT-PCR System utilizes a combination of superior enzymes. The SuperScript IV Reverse Transcriptase mix helps produce an efficient reverse transcription step through its high thermostability, processivity, and ability to synthesize cDNA from variety of RNA samples, while its high sensitivity helps ensure reliable target detection even when RNA input is limited (down to 0.01 pg RNA).

The 2X UniPrime RT-PCR master mix contains modified Platinum SuperFi II DNA Polymerase, which delivers high specificity and high yields and is ideally suited for applications that require sequence accuracy. This novel master mix formulation enables universal primer annealing at 60°C for most primer pairs designed following the general design rules and helps minimize optimization steps, save time, and avoid mistakes in RT-PCR runs.

Two-phase hot start
The innovative two-phase hot-start mechanism helps deliver high specificity and yield in one-step RT-PCR through temporal separation of the activities of the reverse transcriptase and DNA polymerase. This not only allows reaction setup at room temperature, but also helps ensure the stability of assembled reactions for up to 24 hours for targets up to 3 kb and for up to 4h for targets >3 kb.

Colored format
The SuperScript IV UniPrime One-Step RT-PCR System is availabe in a standard dye-free format or colored format containing tracking dyes. When the red tracking dye in the SuperScript IV RT mix and the blue tracking dye in the UniPrime RT-PCR master mix are combined during reaction setup, the final reaction mix turns to purple. This allows for simple visual tracking of reaction setup and helps prevent pipetting errors. The final reaction product can also be directly loaded on agarose gels for a more streamlined workflow.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • SuperScript IV RT Mix, 50 μL
• 2X UniPrime RT-PCR Master Mix, 625 μL
• Nuclease-free water, 1.25 mL

Store at -15 to -25°C.

Description Dye-free
Detection Method Gel Electrophoresis
Format 1-step RT-PCR Kit
GC-Rich PCR Performance High
PCR Method 1-step RT-PCR
Technique 1-step RT-PCR
Multiplex Capability Yes
Optimal Reaction Temperature 50°C to 65°C
Quantity 25 reactions
Reverse Transcriptase SuperScript IV
Hot Start Built-in Hot Start
No. of Reactions 25 x 50 μL Reactions
Reaction Format Master Mix
Reagent Type Reverse Transcription
Size (Final Product) Up to 13 kb
Starting Material RNA
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.