Learn More
Thermo Scientific™ SuperSealer
Description
Ensure a proper seal on plates, blocks and storage tube racks with the Thermo Scientific™ SuperSealer. Choose individual sealing options such as Thermo Scientific™ Matrix™ SepraSeal mats for liquid handling and single tube accessibility, or seal entire plates and blocks for storage with Thermo Scientific™ Nunc™ CapMats. Effectively apply adhesive tape seals and ensure a tight seal with the included SuperSealer Adhesive Seal Mat.
Sealing Simplicity
- Simplifies the sealing of plates, blocks and tube racks of virtually any height, with no adjustments needed, ensuring a secure and consistent seal
- Once the seal along with the plate, block or tube rack are placed into the drawer, simply close the door to initiate sealing
Sealing Flexibility
- Designed to work with Thermo Scientific™ Matrix™ SepraSeal mats, CapMats and, with the SuperSealer AdhesiveSeal Mat, any pre-cut adhesive tape seal, you can be assured a proper seal regardless of sealing option
Includes: Automatic cap, tape and SepraSeal mat applicator
Specifications
Specifications
Description | Thermo Scientific Super Sealer |
No. per Pack | 1 Ea. |
Product Type | Microplate Sealer |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.