Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SuperSignal™ West Pico PLUS Chemiluminescent Substrate

Catalog No. pi34579
Click to view available options
Quantity:
20 mL
200 mL
500 mL
1 L
4 x 500 mL
4 x 1 L

Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) horseradish peroxidase (HRP) substrate that enables picogram to high femtogram-level protein detection by western blot analysis.

Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) horseradish peroxidase (HRP) substrate that enables picogram to high femtogram-level protein detection by western blot analysis.

SuperSignal West Pico PLUS Chemiluminescent Substrate characteristics include:

  • Sensitivity: pictogram to femtogram sensitivity
  • Stability: 8-hr working solution stability; 1-year kit stability at room temperature
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: 6 to 24 hours
  • Recommended primary antibody concentration: 1:1000–1:5000 dilution (0.2–1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:20,000–100,000 dilution (10–50 ng/mL)

SuperSignal West Pico PLUS substrate is designed to provide excellent signal intensity and sensitivity for western blotting with HRP conjugates. The intensity of the light emission combined with the exceptional duration allows for the acquisition of multiple exposures to more easily obtain publication-quality blot images. SuperSignal West Pico PLUS substrate is compatible with different membranes, blocking reagents, and a wide range of antibody dilutions, making it an ideal choice for most western blotting applications.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Substrate
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line SuperSignal™
Quantity 20 mL
Conjugate HRP
Substrate Horseradish Peroxidase
Recommended Antibody Concentrations 2°: 1:20-100 K (10-50 ng/mL), 1°: 1:1 K (0.2-1 μg/mL)
Sensitivity Picogram to high femtogram-level
Signal Duration 6 to 24 hr.
Sufficient For 2 Mini-Gel Blots, 200 cm2 of Membrane
Kit Contents Luminol/Enhancer, 10 mL, Stable Peroxide Buffer, 10 mL
Content And Storage Sufficient for 2 mini-gel blots; 200 cm2 of membrane
• SuperSignal West Pico PLUS Luminol/Enhancer, 10 mL
• SuperSignal West Pico PLUS Stable Peroxide, 10 mL

Store at room temperature.
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.