Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SuperSignal™ Western Blot Enhancer

Catalog No. p-4532085
Click to view available options
Quantity:
50 mL
500 mL

Increase both signal intensity and sensitivity 3- to 10-fold with this membrane treatment reagent and primary antibody diluent.

Thermo Scientific™ SuperSignal Western Blot Enhancer contains a membrane treatment reagent and a primary antibody diluent that increase both signal intensity and sensitivity 3- to 10-fold compared to detection performed without it.

When a protein or antigen is difficult to detect because of low abundance or poor immunoreactivity, use of SuperSignal Western Blot Enhancer can significantly reduce background and enhance detection of low-abundance and weakly immunoreactive antigens. This versatile kit works with PVDF and nitrocellulose membranes and is compatible with fluorescent, chromogenic and chemiluminescent detection.

Highlights:

  • Increase sensitivity – achieve 3- to 10-fold increase in signal intensity and sensitivity

  • Improve specificity – improves signal-to-noise ratio for poor quality and low affinity antibodies

  • Better clarity – reduces background for cleaner Western blots

  • Membrane compatibility – provides effective signal enhancement with PVDF and nitrocellulose membranes

  • Substrate compatibility – validated for use with chromogenic, chemiluminescent and fluorescent detection methods

TRUSTED_SUSTAINABILITY

Specifications

Product Type Western Blot Enhancer
Detection Method Chemiluminescence, Fluorescence, Colorimetric
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line SuperSignal™
Quantity 50 mL
Content And Storage Upon receipt store solutions at 4°C Products are shipped at ambient temperature.
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.