Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Stainless-Steel Micro-Spatulas
GREENER_CHOICESCI_ED

Catalog No. s09578 Shop All Supertek Scientific Products
Change view
Click to view available options
Length (Metric):
20 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S09578 20 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S09578 Supplier Supertek™ Supplier No. CH12221A
Only null left
Add to Cart
Add to Cart

With thin flexible rounded ends.

  • Length: 20 cm (7.87 in.)
  • Sold in packs of 12

Specifications

Autoclavable Autoclavable
Shape Round End
Product Type Micro Spatula
Length (English) 7.87 in.
Includes Thin flexible rounded ends
Material Stainless Steel
Length (Metric) 20 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.