Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Suppressor of Ty 4 homolog 1 Antibody (OTI6B10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP274412A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274412A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274412A30 Supplier Novus Biologicals Supplier No. NBP274412AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suppressor of Ty 4 homolog 1 Monoclonal specifically detects Suppressor of Ty 4 homolog 1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Suppressor of Ty 4 homolog 1
Applications Western Blot
Classification Monoclonal
Clone OTI6B10
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DRB sensitivity-inducing factor 14 kDa subunit, DRB sensitivity-inducing factor small subunit, DSIF p14, DSIF small subunit, SPT4, SPT4HhSPT4, suppressor of Ty (S.cerevisiae) 4 homolog 1, suppressor of Ty 4 homolog 1 (S. cerevisiae), SUPT4H, transcription elongation factor SPT4
Gene Symbols SUPT4H1
Host Species Mouse
Immunogen Full length recombinant protein of human Suppressor of Ty 4 homolog 1 (NP_003159) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6827.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.