Learn More
Bio-Serv™ Supreme Mini-Treats™ Round Nutritional Pellets, Certified

Nutritional mini-treat that comes in different flavors is a good source of calcium and can be used as a reward for doing a good job
$48.75 - $155.78
Specifications
Product Type | Nutritional Pellets |
---|---|
Calories | 3.46 |
Components | Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corn Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Natural and Artificial Flavors, Vitamin Mix, Calcium Silicate, Magnesium Stearate, L-Cystine, Ascorbic Acid, Food Dye (Very Berry Flavor Only) |
Format | Round |
For Use With (Application) | Training, conditioning |
Catalog Number | Mfr. No. | Flavor | Sterility | Size | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Flavor | Sterility | Size | Quantity | Price | Quantity & Availability | ||
14-726-355
![]() ![]() |
Bio-Serv™
F05478 |
Banana | Non-sterile | 1 g | 5000/box |
Each for $155.78
|
|
||
14-726-354
![]() ![]() |
Bio-Serv™
F05475 |
Pina Colada | Non-sterile | 1 g | 5000/box |
Each for $155.78
|
|
||
14-726-356
![]() ![]() |
Bio-Serv™
F05711 |
Very Berry | Non-sterile | 1 g | 5000/box |
Each for $155.78
|
|
||
14-726-342
![]() ![]() |
Bio-Serv™
F054781 |
Banana | Non-sterile | 1 g | 1000/bag |
Each for $48.75
|
|
||
14-726-352
![]() ![]() |
Bio-Serv™
F05472 |
Chocolate | Non-sterile | 1 g | 5000/box |
Each for $155.78
|
|
||
14-726-353
![]() ![]() |
Bio-Serv™
F05474 |
Marshmallow | Non-sterile | 1 g | 5000/box |
Each for $155.78
|
|
||
14-726-346
![]() ![]() |
Bio-Serv™
F057111 |
Very Berry | Non-sterile | 1 g | 1000/bag |
Each for $48.75
|
|
||
14-726-344
![]() ![]() |
Bio-Serv™
F054741 |
Marshmallow | Non-sterile | 1 g | 1000/bag |
Each for $48.75
|
|
||
14-726-343
![]() ![]() |
Bio-Serv™
F054721 |
Chocolate | Non-sterile | 1 g | 1000/bag |
Each for $48.75
|
|
||
14-726-345
![]() ![]() |
Bio-Serv™
F054751 |
Pina Colada | Non-sterile | 1 g | 1000/bag |
Each for $48.75
|
|
||
Description
- Appetizing aroma helps to deter rodent maternal cannibalism; chocolate and very berry flavor recommended
- Excellent for training, conditioning or as a reward following a less desirable procedure
- Nutritionally complete 1gm treat that compliments enrichment programs
- Diameter: 11.43mm (0.45 in.)
- Excellent source of vitamin C
- Certified (Contaminant Screened)
- Nutritionally assayed
- Shape: pellet
- Weight: 1gm
For training/conditioning
Certifications
Contaminant Screened
Specifications
Nutritional Pellets | |
Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corn Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Natural and Artificial Flavors, Vitamin Mix, Calcium Silicate, Magnesium Stearate, L-Cystine, Ascorbic Acid, Food Dye (Very Berry Flavor Only) | |
Training, conditioning | |
1 g | |
Cool dry place | |
0.114 cm |
3.46 | |
Round | |
Pellet | |
Non-sterile | |
0.45 in. |
Safety and Handling
ShelfLife : 1 Year
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.