Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUR-8 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB324999 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB324999 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB324999 Supplier Novus Biologicals Supplier No. NBP299515PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SUR-8 Polyclonal antibody specifically detects SUR-8 in Human samples. It is validated for Western Blot

Specifications

Antigen SUR-8
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias KIAA0862FLJ60412, Protein soc-2 homolog, Protein sur-8 homolog, SIAA0862, SOC-2, soc-2 (suppressor of clear, C.elegans) homolog, soc-2 suppressor of clear homolog (C. elegans), SOC2SUR8leucine-rich repeat protein SHOC-2, SUR-8
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SUR-8.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8036
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.