Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUR1 Antibody (S289-16), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP259320G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259320G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259320G Supplier Novus Biologicals Supplier No. NBP259320G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SUR1 Monoclonal specifically detects SUR1 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SUR1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S289-16
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ABC36, ATP-binding cassette, sub-family C (CFTR/MRP), member 8, HHF1Sulfonylurea receptor 1, HI, PHHIHRINS, sulfonylurea receptor (hyperinsulinemia), SUR1MRP8, SURATP-binding cassette transporter sub-family C member 8, TNDM2ATP-binding cassette sub-family C member 8
Gene Symbols ABCC8
Host Species Mouse
Immunogen Fusion protein amino acids 1548-1582 (cytoplasmic C-terminus) of rat SUR1
Purification Method Protein G purified
Quantity 100 μL
Research Discipline ABC Transporters, Lipid and Metabolism, Nutrient Sensing in the Brain
Primary or Secondary Primary
Gene ID (Entrez) 6833
Test Specificity Detects 160kDa. Does not cross-react with SUR2B.
Target Species Human, Mouse, Rat, Hamster
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.