Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SURF4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP144374 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144374 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144374 Supplier Novus Biologicals Supplier No. NBP144374
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SURF4 Polyclonal specifically detects SURF4 in Human, Mouse, Rat, Bovine, Chicken, Xenopus samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SURF4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS with 0.1% Sodium Azide
Gene Alias ERV29, FLJ22993, MGC102753, SURF-4, surface 4 integral membrane protein, surfeit 4, surfeit locus protein 4
Gene Symbols SURF4
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from N-Terminus of human SURF4.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6836
Test Specificity This product is specific for Surfeit 4 (SURF4). Unless other species reactivities are specified there is no testing data currently available.
Target Species Human, Mouse, Rat, Bovine, Chicken, Xenopus species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.