Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SURF5 Antibody (PCRP-MED22-1E4), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB156288 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB156288 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB156288 Supplier Novus Biologicals Supplier No. NBP314190AF488
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SURF5 Monoclonal antibody specifically detects SURF5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen SURF5
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-MED22-1E4
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Med24, mediator complex subunit 22SURF5surfeit 5, mediator of RNA polymerase II transcription subunit 22, MGC48682, Surf-5, Surfeit locus protein 5
Host Species Mouse
Immunogen Recombinant full-length human SURF5 protein (Uniprot: Q15528)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6837
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.