Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surface Lipoprotein p27 Antibody, Novus Biologicals™
SDP

Catalog No. NBP221652 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221652 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221652 Supplier Novus Biologicals Supplier No. NBP221652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Surface Lipoprotein p27 Polyclonal specifically detects Surface Lipoprotein p27 in Bacteria samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Surface Lipoprotein p27
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:1000
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. O50951
Gene Alias BB_A60, Surface lipoprotein P27
Gene Symbols BB_A60
Host Species Rabbit
Immunogen MBP-fusion protein corresponding to Borrelia burgdorferi Surface Lipoprotein p27 protein. (Uniprot: O50951)
Molecular Weight of Antigen 32.44 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1194336
Test Specificity It is directed against, and shows specific reactivity for, Borrelia burgdorferi Surface Lipoprotein p27 protein. Reactivity with Surface Lipoprotein p27 protein from other sources has not been determined.
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Bacteria
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.