Learn More
United Scientific™ Surface Plasmon Resonance Apparatus
Shop All United Scientific Supplies, Inc Products
Click to view available options
:
Surface Plasmon Resonance Apparatus
Description
- Use a light beam to excite resonant electron oscillations under specific conditions.
- Includes a collimated laser, polarizer, and photodetector.
- Surface Plasmon Resonance is an analytical technique used for detecting and tracking a wide variety of surface processes such as DNA and protein interactions, drug mechanisms, immunoassays, anodic stripping, corrosion, gas detection, and many more.
- Requires (not included) an intermediate spectrometer (SPECT02).
Specifications
Specifications
| Product Type | Optics Set |
| Shape | Various |