Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC alpha Antibody (MC5), Novus Biologicals™
SDP

Catalog No. NB600201SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600201SS 0.025 mL
NB600201 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600201SS Supplier Novus Biologicals Supplier No. NB600201SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 19 publications

PKC alpha Monoclonal specifically detects PKC alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Fish, Baboon, Monkey, Rabbit, Sheep, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Radioimmune Assays.

Specifications

Antigen PKC alpha
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone MC5
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:200-1:500, Immunohistochemistry-Frozen 1:200-1:500, Radioimmunoassay
Gene Accession No. P17252, P04409
Gene Alias AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha
Gene Symbols PRKCA
Host Species Mouse
Immunogen Purified bovine brain Protein Kinase C
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5578
Test Specificity This antibody is capable of detecting PKC beta as well.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Fish, Baboon, Monkey, Rabbit, Sheep, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a Kappa
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.