Learn More
Description
Specifications
Specifications
| Antigen | Survivin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Simple Western 1:25, Flow Cytometry reported in scientific literature (PMID 17875988; 33737139), ELISA reported in scientific literature (PMID 24102797), Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 12671708), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated, Dual RNAscope ISH-IHC |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | O15392 |
| Gene Alias | API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha |
| Gene Symbols | BIRC5 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
