Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Survivin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500201SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500201SS 0.025 mL
NB500201 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB500201SS Supplier Novus Biologicals Supplier No. NB500201SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 341 publication

Survivin Polyclonal specifically detects Survivin in Human, Mouse, Rat, Canine, Feline, Guinea Pig, Hamster samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated, Dual RNAscope ISH-IHC.

Specifications

Antigen Survivin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:25, Flow Cytometry reported in scientific literature (PMID 17875988; 33737139), ELISA reported in scientific literature (PMID 24102797), Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 12671708), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated, Dual RNAscope ISH-IHC
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. O15392
Gene Alias API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
Gene Symbols BIRC5
Host Species Rabbit
Immunogen This Survivin Antibody was developed against full length recombinant human Survivin [UniProt# O15392]
Molecular Weight of Antigen 16 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 332
Target Species Human, Mouse, Rat, Canine, Feline, Guinea Pig, Hamster
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.