Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Survivin Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB001158 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001158 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001158 Supplier Novus Biologicals Supplier No. NB500201R0.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Survivin Polyclonal specifically detects Survivin in Human, Mouse, Rat, Canine, Feline, Guinea Pig, Hamster samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.

Specifications

Antigen Survivin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM Sodium Borate
Gene Alias API4 survivin variant 3 alpha, Apoptosis inhibitor 4 baculoviral IAP repeat-containing protein 5, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, BIRC5, EPR-1, IAP4
Gene Symbols BIRC5
Host Species Rabbit
Immunogen Full length recombinant human Survivin [UniProt# O15392]
Purification Method Affinity Purified
Quantity 0.1 mL
Gene ID (Entrez) 332
Target Species Human, Mouse, Rat, Canine, Feline, Guinea Pig, Hamster
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.