Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Survivin, p Thr34 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB50236A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50236A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50236A532 Supplier Novus Biologicals Supplier No. NB500236AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Survivin Polyclonal specifically detects Survivin in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Survivin
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
Gene Symbols BIRC5
Host Species Rabbit
Immunogen This Survivin [p Thr34] Antibody was developed against a synthetic peptide with a phosphorylated Threonine (amino acid 34) corresponding to human Survivin. [UniProt# O15392]
Molecular Weight of Antigen 16 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 332.0
Test Specificity Survivin [p Thr34] Antibody [NB500-236] is specific for phosphorylated survivin.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.