Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Survivin Rabbit Polyclonal Antibody with BSA and azide

Survivin, Rabbit Polyclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Specifications

Antigen Survivin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RB9245P0
View Documents
Epredia™
RB9245P0
Concentrated 100 μL N/A N/A N/A
RB9245P1
View Documents
Epredia™
RB9245P1
Concentrated 500 μL N/A N/A N/A
RB9245P
View Documents
Epredia™
RB9245P
Concentrated 1 mL N/A N/A N/A
RB9245R1
View Documents
Epredia™
RB9245R1
Prediluted 1 mL N/A N/A N/A
RB9245R7
View Documents
Epredia™
RB9245R7
Prediluted 7 mL N/A N/A N/A
Description

Description

Survivin is a new inhibitor of apoptosis (IAP) protein. It is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. The overexpression of surviving in cancer may overcome this apoptotic checkpoint and favor aberrant progression of transformed cells through mitosis.

Host Species: Rabbit

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of human survivin

Molecular Weight: 16.5kDa

Positive Control: Normal pancreas (α-cells) or prostate CA

Cellular Localization: Cytoplasmic and Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Survivin
Polyclonal
Rabbit
Apoptosis
A synthetic peptide derived from the C-terminus of human survivin
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.