Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUSD1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB413799 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB413799 0.025 mg
NBP261129 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB413799 Supplier Novus Biologicals Supplier No. NBP2611290.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SUSD1 Polyclonal specifically detects SUSD1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SUSD1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias RP11-4O1.1, sushi domain containing 1
Gene Symbols SUSD1
Host Species Rabbit
Immunogen SUSD1 antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human SUSD1. Amino Acid Squence: LRITSEWNKVRRHS
Molecular Weight of Antigen 82 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64420
Test Specificity SUSD1 Antibody is predicted to not cross-react with other SUSDfamiliy members.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.