Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUSD2 Antibody (1279B), Alexa Fluor™ 700, R&D Systems™
SDP

Catalog No. MAB90564AFE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB90564AFE 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB90564AFE Supplier Novus Biologicals Supplier No. MAB90564AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SUSD2 Monoclonal specifically detects SUSD2 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blot.

Specifications

Antigen SUSD2
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1279B
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2
Gene Symbols SUSD2
Host Species Rabbit
Immunogen Partial recombinant human SUSD2 protein (amino acids 1-785) [UniProt Q9UGT4]
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56241
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.