Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUSD2 Antibody (1279B) - Azide and BSA Free, R&D Systems™
SDP

Catalog No. MAB90564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB90564 100 μg
MAB90564SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB90564 Supplier Novus Biologicals Supplier No. MAB90564
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SUSD2 Monoclonal specifically detects SUSD2 in Human, Rat samples. It is validated for CyTOF, Flow Cytometry, ICC/IF (Negative), Immunohistochemistry (Negative), Western Blot.

Specifications

Antigen SUSD2
Applications CyTOF, Flow Cytometry, ICC/IF (Negative), Immunohistochemistry (Negative), Western Blot
Classification Monoclonal
Clone 1279B
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry (Negative), ICC/IF (Negative), CyTOF-ready
Formulation PBS with No Preservative
Gene Alias BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2
Gene Symbols SUSD2
Host Species Rabbit
Immunogen Partial recombinant human SUSD2 protein (amino acids 1-785) [UniProt Q9UGT4]
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 56241
Target Species Human, Rat, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.