Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUSD2 Antibody (1279D), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB310588 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310588 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310588 Supplier Novus Biologicals Supplier No. FAB90563PECY55
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SUSD2 Monoclonal specifically detects SUSD2 in Human, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen SUSD2
Applications Flow Cytometry
Classification Monoclonal
Clone 1279D
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2
Host Species Rabbit
Immunogen HEK293 human embryonic kidney cell line-derived human SUSD2, Met1-Ala785, Accession # Q9UGT4
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 56241
Target Species Human, Rat
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.