Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUV39H1 Monoclonal Antibody (MG44), Active Motif™

Mouse Monoclonal Antibody
$156.65 - $638.65
Specifications
Antigen | SUV39H1 |
---|---|
Clone | MG44 |
Concentration | 1 mg/mL |
Applications | ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2851
![]() ![]() |
Active Motif
39786 |
10 μg |
Each for $156.65
|
|
|||||
50-199-2850
![]() ![]() |
Active Motif
39785 |
100 μg |
Each for $638.65
|
|
|||||
Description
This gene encodes an evolutionarily-conserved protein containing an N-terminal chromodomain and a C-terminal SET domain. The encoded protein is a histone methyltransferase that trimethylates lysine 9 of histone H3, which results in transcriptional gene silencing. Loss of function of this gene disrupts heterochromatin formation and may cause chromosome instability. Alternative splicing results in multiple transcript variants.Specifications
SUV39H1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
AI852103; AL022883; DXHXS7466e; H3-K9-HMTase 1; Histone H3-K9 methyltransferase 1; Histone-lysine N-methyltransferase SUV39H1; histone-lysine N-methyltransferase, H3 lysine-9 specific 1; KMT1A; Lysine N-methyltransferase 1A; MG44; mIS6; Position-effect variegation 3-9 homolog; RGD1565028; Su(var)3-9 homolog 1; Suppressor of variegation 3-9 homolog 1; suppressor of variegation 3-9 homolog 1 (Drosophila); SUV39H; Suv39h histone methyltransferase; SUV39H1 | |
SUV39H1 | |
IgG1 | |
Protein G | |
Antibody |
MG44 | |
ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
O43463, O54864 | |
20937, 6839 | |
This SUV39H1 antibody was raised against a recombinant protein corresponding to amino acids 1-126 of mouse SUV39H1. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
SUV39H1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title